COMH42116DE1 Combi-Blocks, Inc.

1,4-Phenylene-4,4'-bis(2,6-diphenyl-4-pyrylium tetrafluoroborate)

  • MDL number MFCD00160234
  • Cas Number : 54620-09-6
  • Molecular Weight : 714.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1052

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1052
Availability details for
Quantity
  • Availability
  • Add to Cart