COMH42116DE5 Combi-Blocks, Inc.

(Pentylsulfanyl)methanimidamide hydrobromide

  • MDL number MFCD01694539
  • Cas Number : 4270-00-2
  • Molecular Weight : 227.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name pentyl carbamimidothioate hydrobromide
Inchi Code 1S/C6H14N2S.BrH/c1-2-3-4-5-9-6(7)8;/h2-5H2,1H3,(H3,7,8);1H
Inchi Key JTEBEBGEFYAPTQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1064

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1064
Availability details for
Quantity
  • Availability
  • Add to Cart