COMH42116DE8 Combi-Blocks, Inc.

(2-Chloro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride

  • MDL number MFCD18380004
  • Cas Number : 1261230-82-3
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1075
Availability details for
Quantity
  • Availability
  • Add to Cart