COMH42116DEB Combi-Blocks, Inc.

(4-Formyl-2,6-dimethylphenol)-m-peg8-acid ester

  • MDL number MFCD28142472
  • Molecular Weight : 544.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1085
Availability details for
Quantity
  • Availability
  • Add to Cart