COMH42116DEC Combi-Blocks, Inc.

Trisulfo-cy3-alkyne

  • MDL number MFCD29052183
  • Cas Number : 1895849-34-9
  • Molecular Weight : 761.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1086
Availability details for
Quantity
  • Availability
  • Add to Cart