COMH42116DF3 Combi-Blocks, Inc.

M-Peg5-2-methylacrylate

  • MDL number MFCD31561108
  • Cas Number : 48074-75-5
  • Molecular Weight : 320.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1121
Availability details for
Quantity
  • Availability
  • Add to Cart