COMH42116DF7 Combi-Blocks, Inc.

6-Nitro-4-oxohexanoic acid

  • MDL number MFCD01464036
  • Cas Number : 98196-97-5
  • Molecular Weight : 175.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1127
Availability details for
Quantity
  • Availability
  • Add to Cart