COMH42116DF9 Combi-Blocks, Inc.

5-(Dimethoxymethyl)-2-fluorobenzonitrile

  • MDL number MFCD31583421
  • Cas Number : 2197045-88-6
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1133
Availability details for
Quantity
  • Availability
  • Add to Cart