COMH42116E01 Combi-Blocks, Inc.

(2E)-3-(Dimethylamino)-1-(4-ethoxyphenyl)prop-2-en-1-one

  • MDL number MFCD14703339
  • Cas Number : 839726-94-2
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1154
Availability details for
Quantity
  • Availability
  • Add to Cart