COMH42116E08 Combi-Blocks, Inc.

2,2-Dimethyl-tetrahydro-pyran-6-methanol

  • MDL number MFCD26131217
  • Cas Number : 1849358-51-5
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1171
Availability details for
Quantity
  • Availability
  • Add to Cart