COMH42116E0C Combi-Blocks, Inc.

4-Amino-1h-pyrazole-5-carboxamide dihydrochloride

  • MDL number MFCD16038566
  • Molecular Weight : 199.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1202

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1202
Availability details for
Quantity
  • Availability
  • Add to Cart