COMH42116E18 Combi-Blocks, Inc.

4-Formyl-1-(4-methoxybenzyl)-2,5-dimethyl-1h-pyrrole-3-carboxylic acid

  • MDL number MFCD14281672
  • Cas Number : 1171868-54-4
  • Molecular Weight : 287.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1241

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1241
Availability details for
Quantity
  • Availability
  • Add to Cart