COMH42116E2A Combi-Blocks, Inc.

Boc-l-met-o-ch2-f-ch2-cooh

  • MDL number MFCD00273507
  • Cas Number : 66271-05-4
  • Molecular Weight : 397.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1321
Availability details for
Quantity
  • Availability
  • Add to Cart