COMH42116E2E Combi-Blocks, Inc.

4-Methyloxazole-5-acetic acid

  • MDL number MFCD19230094
  • Cas Number : 98140-74-0
  • Molecular Weight : 141.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1330
Availability details for
Quantity
  • Availability
  • Add to Cart