COMH42116E40 Combi-Blocks, Inc.

5-(5-Trifluoromethyl-furan-2-ylmethylsulfanyl)-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD28248746
  • Cas Number : 1993122-63-6
  • Molecular Weight : 281.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(((5-(trifluoromethyl)furan-2-yl)methyl)thio)-1,3,4-thiadiazol-2-amine
Inchi Code 1S/C8H6F3N3OS2/c9-8(10,11)5-2-1-4(15-5)3-16-7-14-13-6(12)17-7/h1-2H,3H2,(H2,12,13)
Inchi Key PUAHPRLSWLGTSI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1408

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1408
Availability details for
Quantity
  • Availability
  • Add to Cart