COMH42116E4E Combi-Blocks, Inc.

2-(Aminomethyl)-3,5-dimethyl-4h-1,2'-bipyridin-4-one dihydrochloride

  • MDL number MFCD31619052
  • Cas Number : 2279121-93-4
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1471
Availability details for
Quantity
  • Availability
  • Add to Cart