COMH42116E52 Combi-Blocks, Inc.

3-(Dimethylamino)-5-phenylisoxazole-4-carboxylic acid

  • MDL number MFCD31583404
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1487
Availability details for
Quantity
  • Availability
  • Add to Cart