COMH42116E5B Combi-Blocks, Inc.

2-(Diethylamino)-2-phenylacetonitrile

  • MDL number MFCD01673030
  • Cas Number : 5097-99-4
  • Molecular Weight : 188.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1542
Availability details for
Quantity
  • Availability
  • Add to Cart