COMH42116E73 Combi-Blocks, Inc.

(3S,3Ar,5s,6as,7s)-octahydro-3,5-methanocyclopenta[b]pyrrol-7-ylmethanol

  • MDL number MFCD31614359
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1680
Availability details for
Quantity
  • Availability
  • Add to Cart