COMH42116E74 Combi-Blocks, Inc.

Methyl 5-(4-chlorophenyl)-3-nitroisoxazole-4-carboxylate

  • MDL number MFCD31583413
  • Cas Number : 1818225-38-5
  • Molecular Weight : 282.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1687
Availability details for
Quantity
  • Availability
  • Add to Cart