COMH42116E77 Combi-Blocks, Inc.

5-(Benzylamino)pentanoic acid hydrochloride

  • MDL number MFCD31557153
  • Cas Number : 137565-39-0
  • Molecular Weight : 243.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(benzylamino)pentanoic acid hydrochloride
Inchi Code 1S/C12H17NO2.ClH/c14-12(15)8-4-5-9-13-10-11-6-2-1-3-7-11;/h1-3,6-7,13H,4-5,8-10H2,(H,14,15);1H
Inchi Key SRPVDAAVDABCPI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1707
Availability details for
Quantity
  • Availability
  • Add to Cart