COMH42116E81 Combi-Blocks, Inc.

Trisulfo-cy5.5-alkyne

  • MDL number MFCD29052181
  • Cas Number : 2055046-12-1
  • Molecular Weight : 954.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-((1E,3E,5E)-5-(3-ethyl-1,1-dimethyl-6,8-disulfo-1,3-dihydro-2H-benzo[e]indol-2-ylidene)penta-1,3-dien-1-yl)-1,1-dimethyl-3-(6-oxo-6-(prop-2-yn-1-ylamino)hexyl)-8-sulfo-1H-benzo[e]indol-3-ium-6-sulfonate
Inchi Code 1S/C44H47N3O13S4/c1-7-22-45-40(48)17-13-10-14-23-47-35-21-19-31-33(25-29(62(52,53)54)27-37(31)64(58,59)60)42(35)44(5,6)39(47)16-12-9-11-15-38-43(3,4)41-32-24-28(61(49,50)51)26-36(63(55,56)57)30(32)18-20-34(41)46(38)8-2/h1,9,11-12,15-16,18-21,24-27H,8,10,13-14,17,22-23H2,2-6H3,(H4-,45,48,49,50,51,52,53,54,55,56,57,58,59,60)
Inchi Key ORXCITITVUDFJM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1742
Availability details for
Quantity
  • Availability
  • Add to Cart