COMH42116E84 Combi-Blocks, Inc.

Fmoc-peg3-ala-ala-asn(trt)-pab

  • MDL number MFCD30182025
  • Cas Number : 2055042-69-6
  • Molecular Weight : 1047.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1751

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1751
Availability details for
Quantity
  • Availability
  • Add to Cart