COMH42116E87 Combi-Blocks, Inc.

1-(5-Nitropyridin-2-yl)piperidin-4-amine dihydrochloride

  • MDL number MFCD27756350
  • Cas Number : 412355-82-9
  • Molecular Weight : 295.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C10H14N4O2.2ClH/c11-8-3-5-13(6-4-8)10-2-1-9(7-12-10)14(15)16;;/h1-2,7-8H,3-6,11H2;2*1H
Inchi Key JOCRHSZWXXKTMA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1775
Availability details for
Quantity
  • Availability
  • Add to Cart