COMH42116EA0 Combi-Blocks, Inc.

Ethyl 2-[(2,4-difluorophenyl)amino]-4-methylpyrimidine-5-carboxylate

  • MDL number MFCD31581340
  • Cas Number : 2270909-91-4
  • Molecular Weight : 293.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1947
Availability details for
Quantity
  • Availability
  • Add to Cart