COMH42116EB3 Combi-Blocks, Inc.

4-(3-Fluorobenzyl)-5-methyl-1,2-dihydro-3h-pyrazol-3-one

  • MDL number MFCD31581344
  • Cas Number : 2270908-45-5
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2026
Availability details for
Quantity
  • Availability
  • Add to Cart