COMH42116EB4 Combi-Blocks, Inc.

3-Amino-3-pyridin-3-ylpropanoic acid hydrochloride

  • MDL number MFCD27756356
  • Cas Number : 1983937-54-7
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C8H10N2O2.ClH/c9-7(4-8(11)12)6-2-1-3-10-5-6;/h1-3,5,7H,4,9H2,(H,11,12);1H
Inchi Key KEHMWCRFVPPZEO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2031
Availability details for
Quantity
  • Availability
  • Add to Cart