COMH42116EC9 Combi-Blocks, Inc.

Pc alkyne-peg4-nhs carbonate ester

  • MDL number MFCD29052184
  • Cas Number : 1802907-98-7
  • Molecular Weight : 653.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2193

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2193
Availability details for
Quantity
  • Availability
  • Add to Cart