COMH42116ECA Combi-Blocks, Inc.

3-Bromo-1-(4-ethylphenyl)-1h-pyrrole-2,5-dione

  • MDL number MFCD31581332
  • Cas Number : 2186672-64-8
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2195
Availability details for
Quantity
  • Availability
  • Add to Cart