COMH42116ED9 Combi-Blocks, Inc.

1-(1,4-Dioxan-2-yl)cyclopropanamine hydrochloride

  • MDL number MFCD31557156
  • Cas Number : 2270904-97-5
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2305

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2305
Availability details for
Quantity
  • Availability
  • Add to Cart