COMH42116EDF Combi-Blocks, Inc.

(Z)-2-Bromo-2-butenoic acid

  • MDL number MFCD00154334
  • Cas Number : 5405-34-5
  • Molecular Weight : 165.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2342

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2342
Availability details for
Quantity
  • Availability
  • Add to Cart