COMH42116EE2 Combi-Blocks, Inc.

Propargyl-peg1-ss-peg1-pfp ester

  • MDL number MFCD29042354
  • Cas Number : 1817735-30-0
  • Molecular Weight : 430.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2363

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2363
Availability details for
Quantity
  • Availability
  • Add to Cart