COMH42116EE5 Combi-Blocks, Inc.

tert-Butyl (4ar,7as)-hexahydropyrrolo[3,4-b][1,4]oxazine-6(2h)-carboxylate hydrochloride

  • MDL number MFCD31557136
  • Cas Number : 2365390-56-1
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2399
Availability details for
Quantity
  • Availability
  • Add to Cart