COMH42116EE6 Combi-Blocks, Inc.

5-(1H-1,3-Benzodiazol-2-yl)pentan-1-amine dihydrochloride

  • MDL number MFCD00707897
  • Cas Number : 94276-03-6
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2404
Availability details for
Quantity
  • Availability
  • Add to Cart