COMH42116EE9 Combi-Blocks, Inc.

Methyl 4-[(2-methylbenzoyl)amino]benzoate

  • MDL number MFCD01034018
  • Cas Number : 200278-95-1
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2417
Availability details for
Quantity
  • Availability
  • Add to Cart