COMH42116EED Combi-Blocks, Inc.

(2-(2-[2-(2-Mercapto-ethoxy)-ethoxy]-ethoxy)-ethyl)phosphonic acid

  • MDL number MFCD21596687
  • Cas Number : 1360716-36-4
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2455
Availability details for
Quantity
  • Availability
  • Add to Cart