COMH42116EF0 Combi-Blocks, Inc.

1-(3-Methylbenzyl)-1h-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD16674247
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2465
Availability details for
Quantity
  • Availability
  • Add to Cart