COMH42116EF3 Combi-Blocks, Inc.

7-Amino-3-isobutylquinazolin-4(3h)-one

  • MDL number MFCD31583454
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2479
Availability details for
Quantity
  • Availability
  • Add to Cart