COMH42116EF5 Combi-Blocks, Inc.

Methyl 6-(3-chlorophenyl)-2-oxo-1,2-dihydropyridine-3-carboxylate

  • MDL number MFCD31581342
  • Cas Number : 2120828-58-0
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2503

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2503
Availability details for
Quantity
  • Availability
  • Add to Cart