COMH42116EFF Combi-Blocks, Inc.

1-(Bromomethyl)-4-iodo-2-methoxybenzene

  • MDL number MFCD20924441
  • Cas Number : 1824344-38-8
  • Molecular Weight : 327

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2553
Availability details for
Quantity
  • Availability
  • Add to Cart