COMH42116F04 Combi-Blocks, Inc.

3-[1-(2-Furylmethyl)-2,5-dioxoimidazolidin-4-yl]propanoic acid

  • MDL number MFCD18485940
  • Cas Number : 1214026-62-6
  • Molecular Weight : 252.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2574
Availability details for
Quantity
  • Availability
  • Add to Cart