COMH42116F05 Combi-Blocks, Inc.

3-Pentadecyl-1h-1,2,4-triazol-5-amine

  • MDL number MFCD31619045
  • Cas Number : 101593-19-5
  • Molecular Weight : 294.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2579
Availability details for
Quantity
  • Availability
  • Add to Cart