COMH42116F06 Combi-Blocks, Inc.

N-(3-Aminopropyl)-2-hydroxybenzamide hydrochloride

  • MDL number MFCD31583446
  • Cas Number : 2270906-39-1
  • Molecular Weight : 230.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-aminopropyl)-2-hydroxybenzamide hydrochloride
Inchi Code 1S/C10H14N2O2.ClH/c11-6-3-7-12-10(14)8-4-1-2-5-9(8)13;/h1-2,4-5,13H,3,6-7,11H2,(H,12,14);1H
Inchi Key SEGCYWHXICHBQR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2587
Availability details for
Quantity
  • Availability
  • Add to Cart