COMH42116F08 Combi-Blocks, Inc.

N-(1H-Indol-5-ylmethyl)-n-(2-methoxyethyl)amine

  • MDL number MFCD12197625
  • Cas Number : 946757-24-0
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2595

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2595
Availability details for
Quantity
  • Availability
  • Add to Cart