COMH42116F0A Combi-Blocks, Inc.

4,5-Dimethoxy-2-(1h-pyrrol-1-yl)benzoic acid

  • MDL number MFCD09736022
  • Cas Number : 55540-38-0
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2603
Availability details for
Quantity
  • Availability
  • Add to Cart