COMH42116F0F Combi-Blocks, Inc.

Ethyl n-(4-fluoro-2-hydroxyphenyl)glycinate

  • MDL number MFCD31583430
  • Cas Number : 2270905-44-5
  • Molecular Weight : 213.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (4-fluoro-2-hydroxyphenyl)glycinate
Inchi Code 1S/C10H12FNO3/c1-2-15-10(14)6-12-8-4-3-7(11)5-9(8)13/h3-5,12-13H,2,6H2,1H3
Inchi Key SITUSRPZJDDGTB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2627

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2627
Availability details for
Quantity
  • Availability
  • Add to Cart