COMH42116F10 Combi-Blocks, Inc.

3-Amino-6-hydroxy-2,4-dimethylpyridine dihydrochloride

  • MDL number MFCD09743471
  • Cas Number : 2270908-38-6
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-4,6-dimethyl-2-pyridinol dihydrochloride
Inchi Code 1S/C7H10N2O.2ClH/c1-4-3-6(10)9-5(2)7(4)8;;/h3H,8H2,1-2H3,(H,9,10);2*1H
Inchi Key ZAQBNEFAKGLFGE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2637

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2637
Availability details for
Quantity
  • Availability
  • Add to Cart