COMH42116F13 Combi-Blocks, Inc.

3-[(4-Methoxyphenyl)amino]-1,3-dihydro-2h-indol-2-one

  • MDL number MFCD12030966
  • Cas Number : 1099622-05-5
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2645
Availability details for
Quantity
  • Availability
  • Add to Cart