COMH42116F2A Combi-Blocks, Inc.

Methyltetrazine-sulfo-nhs ester

  • MDL number MFCD29052195
  • Cas Number : 1821017-46-2
  • Molecular Weight : 429.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2749

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2749
Availability details for
Quantity
  • Availability
  • Add to Cart