COMH42116F83 Combi-Blocks, Inc.

Sulfo dbco-peg4-amine

  • MDL number MFCD30458045
  • Cas Number : 2055198-05-3
  • Molecular Weight : 674.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2779
Availability details for
Quantity
  • Availability
  • Add to Cart